Skip to Content
MilliporeSigma
All Photos(1)

Documents

S1299

Supelco

Sucrose assay reagent

for use with enzymatic assay kit SCA20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12164500
NACRES:
NA.84

Quality Level

analyte chemical class(es)

sugars (sucrose)

technique(s)

photometry: suitable

application(s)

food and beverages
general analytical

storage temp.

2-8°C

accessory

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service