PHR8942
Hydromorphone HCl
pharmaceutical secondary standard, certified reference material
Synonym(s):
Hydromorphone hydrochloride, 4,5-epoxy-3-hydroxy-7-methylmorphinan-6-one hydrochloride
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Agency
traceable to USP 1323000
API family
hydromorphone
form
solid
packaging
pkg of 50 mg
application(s)
pharmaceutical
SMILES string
OC1=C(O2)C3=C(C[C@@H]4[C@@]5([H])[C@@]3([C@]2([H])C(CC5)=O)CCN4C)C=C1.[H]Cl
InChI
1S/C17H19NO3.ClH/c1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18;/h2,4,10-11,16,19H,3,5-8H2,1H3;1H/t10-,11+,16-,17-;/m0./s1
InChI key
XHILEZUETWRSHC-NRGUFEMZSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service