Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHR1507

Supelco

2-Amino-5-Chlorobenzophenone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Amino-5-chlorobenzophenone, 4-Chloro-2-benzoylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(Cl)COC6H5
CAS Number:
Molecular Weight:
231.68
Beilstein:
475640
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1022808

description

API family: alprazolam

API family

alprazolam, oxazepam; prazepam, chlordiazepoxide 

CofA

current certificate can be downloaded

packaging

pkg of 200 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

96-98 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccc(Cl)cc1C(=O)c2ccccc2

InChI

1S/C13H10ClNO/c14-10-6-7-12(15)11(8-10)13(16)9-4-2-1-3-5-9/h1-8H,15H2

InChI key

ZUWXHHBROGLWNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3314 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service