Skip to Content
MilliporeSigma
All Photos(2)

Documents

NIST920

D-Mannitol

NIST® SRM® 920

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

NIST®

mp

167-170 °C (lit.)

application(s)

general analytical

format

neat

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for use in the calibration and standardization of procedures for triglyceride determinations in chemical analysis and for the critical evaluation of routine working or secondary reference materials used in these procedures. It is certified as a chemical of known purity. A unit of SRM 920 consists of 50 g of material. For more information, please refer to the SDS and COA.

SRM 920_cert

SRM 920_SDS

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
D-Mannitol

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service