Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3130A

Lutetium (Lu) standard solution

NIST® SRM® 3130a

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 10 ML
$2,070.00

$2,070.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 X 10 ML
$2,070.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,070.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

format

single component solution

General description

This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of lutetium. A unit of SRM 3130a consists of five 10 mL sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically from high-purity lutetium oxide to contain a known mass fraction of lutetium. The solution contains nitric acid at a concentration (molarity) of approximately 1.6 mol/L. For more information, please refer ot the COA and SDS.

SRM 3130A_cert

SRM 3130A_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Lutetium (Lu)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service