Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2779

Gulf of mexico crude oil

NIST® SRM® 2779

Synonym(s):

Crude oil

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 1.2 ML
$1,450.00

$1,450.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 X 1.2 ML
$1,450.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,450.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.2 mL

manufacturer/tradename

NIST®

application(s)

petroleum

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

One unit of SRM 2779 contains five ampoules, with each ampoule containing 1.2 mL of crude oil.


SRM 2779_cert

SRM 2779_SDS

Application

The SRM 2779 is intended for the assessment of analytical methods used to determine selected polycyclic aromatic hydrocarbons (PAHs), hopanes, and steranes within a crude oil matrix.

Features and Benefits

  • Accompanied with certificate documenting NIST-certified mass fraction, coverage factor (k), and uncertainty values for (PAHs) in Gulf of mexico crude oil.
  • The certified values offer Metrological traceability to the SI derived unit for intrinsic viscosity (expressed as milliliters per gram).
  • The reference values for additional compounds in Gulf of Mexico crude oil are presented as mass fractions.
  • The NIST certificate is provided with expiration certificate, storage, handling, and maintenance instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

19.4 °F - (External MSDS)

Flash Point(C)

-7 °C - (External MSDS)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service