Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2777

Weathered Gulf of Mexico Crude Oil in Toluene

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 1.2 ML
$2,860.00

$2,860.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 X 1.2 ML
$2,860.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,860.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

packaging

ampule of 5 × 1.2 mL each

manufacturer/tradename

NIST®

application(s)

petroleum

format

matrix material

General description

This SRM is a naturally weathered Gulf of Mexico crude oil that is diluted in toluene. NIST2777 is intended for use in evaluating analytical methods for the determination of polycyclic aromatic hydrocarbons (PAHs) in weathered crude
oil matrices. All the constituents for which certified and reference values are provided are naturally present in weathered oil. A unit of NIST2777 consists of five ampoules, each containing approximately 1.2mL of crude oil soluton.

Other Notes

Certified for the analytes listed below.
Phenanthrene, 1-Methylphenanthrene, 2-Methylphenanthrene, 3-Methylphenanthrene, 9-Methylphenanthrene, Benzo[ghi]perylene

Reference/Informational value is provided for the following analytes. Dibenzothiophene, Pyrene, 1-Methylpyrene, 4-Methylpyrene, Chrysene, Triphenylene, Benzo[b]fluoranthene, Picene, 17α(H),21α(H)-Hopane

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service