Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST1775

Refractory alloy MP-35-N

NIST® SRM® 1775

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

A single unit of SRM 1775 contains a disk measuring around 35 mm (1.375 in) in diameter and 12.7 mm (0.5 in) thick.For more information, please refer to the SDS and COA.
SRM 1242_cert

SRM 1242_SDS

Features and Benefits

  • NIST Certified Mass Fraction values are metrologically traceable to the SI unit of mass.
  • Information Mass FractionValues for Niobium, Nitrogen, Silicon and Tungsten are reported and expressed as Mass Fraction (%)
  • Reference mass fraction values for various constituents are provided and are metrologically traceable to the SI unit of mass.

Other Notes

Certified for the analytes listed below.
Chromium (Cr), Cobalt (Co), Iron (Fe), Molybdenum (Mo), Nickel (Ni),
Titanium (Ti), Boron (B), Manganese (Mn), Sulfur (S), Vanadium (V)

Reference/Informational value is provided for the following analytes.
Aluminum (Al), Carbon (C), Copper (Cu), Phosphorus (P), Niobium (Nb), Nitrogen (N), Silicon (Si), Tungsten (W)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service