Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1617B

Sulfur in kerosene

NIST® SRM® 1617b, (high level)

Synonym(s):

Kerosene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product NIST1617B is currently not available for purchase. Contact Technical Service

grade

certified reference material

Quality Level

vapor density

4.5 (vs air)

vapor pressure

0.23 mmHg ( 20 °C)

autoignition temp.

442 °F

expl. lim.

5 %

packaging

pkg of 100 mL

manufacturer/tradename

NIST®

technique(s)

XRF: suitable

bp

190-250 °C (lit.)

density

0.8 g/mL at 25 °C (lit.)

application(s)

petroleum

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Certified for the analytes listed below.
Sulphur (S)

Reference/Informational value is provided for the following analytes.

Paraffins, Monocycloparaffins, Dicycloparaffins, Alkylbenzenes, Indanes and Tetralins, Naphthalenes

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service