Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DLW354240

Sigma-Aldrich

Corning® Cell-Tak Cell Tissue Adhesive 1 mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Pricing and availability is not currently available.

biological source

Mytilus edulis (Polyphenolic proteins)

form

liquid (in 5% acetic acid)

mol wt

110-140 kDa

packaging

pkg of 1 mg

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Corning® Cell-Tak cell and tissue adhesive, 1 mg, is used to attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP (fluorinated ethylene propylene) polymer and biological materials.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Swist et al.
Nature communications, 11(1), 4479-4479 (2020-09-10)
The giant protein titin is thought to be required for sarcomeric integrity in mature myocytes, but direct evidence for this hypothesis is limited. Here, we describe a mouse model in which Z-disc-anchored TTN is depleted in adult skeletal muscles. Inactivation
Yong Li et al.
eLife, 9 (2020-12-29)
The giant muscle protein titin is a major contributor to passive force; however, its role in active force generation is unresolved. Here, we use a novel titin-cleavage (TC) mouse model that allows specific and rapid cutting of elastic titin to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service