MilliporeSigma
  • Graded titin cleavage progressively reduces tension and uncovers the source of A-band stability in contracting muscle.
Graded titin cleavage progressively reduces tension and uncovers the source of A-band stability in contracting muscle.
eLife (2020-12-29)
Yong Li, Anthony L Hessel, Andreas Unger, David Ing, Jannik Recker, Franziska Koser, Johanna K Freundt, Wolfgang A Linke
ABSTRACT
The giant muscle protein titin is a major contributor to passive force; however, its role in active force generation is unresolved. Here, we use a novel titin-cleavage (TC) mouse model that allows specific and rapid cutting of elastic titin to quantify how titin-based forces define myocyte ultrastructure and mechanics. We show that under mechanical strain, as TC doubles from heterozygous to homozygous TC muscles, Z-disks become increasingly out of register while passive and active forces are reduced. Interactions of elastic titin with sarcomeric actin filaments are revealed. Strikingly, when titin-cleaved muscles contract, myosin-containing A-bands become split and adjacent myosin filaments move in opposite directions while also shedding myosins. This establishes intact titin filaments as critical force-transmission networks, buffering the forces observed by myosin filaments during contraction. To perform this function, elastic titin must change stiffness or extensible length, unveiling its fundamental role as an activation-dependent spring in contracting muscle.
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Potassium propionate, AldrichCPR
Sigma-Aldrich
Magnesium acetate solution, BioUltra, ~1 M in H2O