Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D2679

Supelco

o-Dianisidine dihydrochloride

for enzymic, spectrophotometric determination, vial of 5 mg

Synonym(s):

o-Dianisidine reagent

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$42.30

$42.30


In StockDetails


Request a Bulk OrderRequest more information

Select a Size

Change View
1 VIAL
$42.30

About This Item

EC Number:
UNSPSC Code:
12164500
NACRES:
NA.84

$42.30


In StockDetails


Request a Bulk OrderRequest more information

grade

for enzymic, spectrophotometric determination

Quality Level

form

solid

packaging

vial of 5 mg

technique(s)

photometry: suitable

application(s)

food and beverages
general analytical

storage temp.

2-8°C

Application

Inidcator substrate for the quantitative determination of glucose in food and other materials utilizing the glucose oxidase-peroxidase-o-dianisidine enzymatic assay.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service