Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C6081

Sigma-Aldrich

9-(2-Carboxy-2-cyanovinyl)julolidine

≥98% (HPLC)

Synonym(s):

9-([E]-2-Carboxy-2-cyanovinyl)julolidine, CCVJ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N2O2
CAS Number:
Molecular Weight:
268.31
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

storage temp.

2-8°C

SMILES string

OC(=O)\C(=C\c1cc2CCCN3CCCc(c1)c23)C#N

InChI

1S/C16H16N2O2/c17-10-14(16(19)20)9-11-7-12-3-1-5-18-6-2-4-13(8-11)15(12)18/h7-9H,1-6H2,(H,19,20)/b14-9+

InChI key

JXENNHTVELFRHV-NTEUORMPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescent probe for binding to proteins etc., quantum yield increases by decreasing free rotation: fluorescent molecular rotor.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Iio et al.
Journal of biochemistry, 113(2), 196-199 (1993-02-01)
1-(2-Hydroxyethyl)-6-[(2,2-dicyano)vinyl]-2,3,4-trihydroquinoli ne (DCQ) is a fluorescent dye and its fluorescence yield is determined by a freedom of intramolecular rotation. When DCQ was added to an actin solution, its fluorescence intensity increased as the actin molecule transformed from G-actin to F-actin.
T Iwaki et al.
Biochemistry, 32(29), 7589-7592 (1993-07-27)
We have prepared monoclonal antibodies for the fluorescent molecular rotors 9-(2-carboxy-2-cyanovinyl)julolidine (CCVJ) and 9-(dicyanovinyl)julolidine (DCVJ). Mouse monoclonal antibody (IgG2b) prepared against CCVJ-conjugated bovine serum albumin strongly bound CCVJ and DCVJ. The CCVJ (or DCVJ) binding to IgG and Fab was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service