Skip to Content
MilliporeSigma
9-(2-Carboxy-2-cyanovinyl)julolidine BioReagent, suitable for fluorescence, ≥97.0% (HPLC)

9-(2-Carboxy-2-cyanovinyl)julolidine

9-(2-Carboxy-2-cyanovinyl)julolidine BioReagent, suitable for fluorescence, ≥97.0% (HPLC)
Synonym(s):
9-([E]-2-Carboxy-2-cyanovinyl)julolidine, CCVJ
Empirical Formula (Hill Notation):
C16H16N2O2
CAS Number:
Molecular Weight:
268.31

Products

Brand
Formula Weight
Physical Form
Product Line
Purity
Showing 1-1 of 1