Skip to Content
MilliporeSigma
All Photos(1)

Documents

99357

Supelco

Sodium ionophore II - cocktail A

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700

product line

Selectophore

Quality Level

form

liquid

composition

2-Nitrophenyl octyl ether, 89.5 wt. % (73732)
Sodium ionophore II, 10.0 wt. % (71733)
Sodium tetraphenylborate, 0.5 wt. % (72018)

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service