Skip to Content
MilliporeSigma
All Photos(1)

Documents

71738

Supelco

Sodium ionophore V

Selectophore, function tested

Synonym(s):

4-Octadecanoyloxymethyl-N,N,N′,N′-tetracyclohexyl-1,2-phenylenedioxydiacetamide, ETH 4120

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H88N2O6
CAS Number:
Molecular Weight:
849.28
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

liquid

quality

function tested

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCc1ccc(OCC(=O)N(C2CCCCC2)C3CCCCC3)c(OCC(=O)N(C4CCCCC4)C5CCCCC5)c1

InChI

1S/C53H88N2O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-28-37-53(58)61-41-44-38-39-49(59-42-51(56)54(45-29-20-16-21-30-45)46-31-22-17-23-32-46)50(40-44)60-43-52(57)55(47-33-24-18-25-34-47)48-35-26-19-27-36-48/h38-40,45-48H,2-37,41-43H2,1H3

InChI key

HSRJLFJSCKIVPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Physical form

solution in 0.5 mL heptane

Other Notes

Very lipophilic sodium-selective ionophore for chemical sensors of long lifetime

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Gehrig et al.
Analytica Chimica Acta, 233, 295-295 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service