Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

937061

Sigma-Aldrich

(t-Bu)PhCPhos Pd G4 ChemBeads

new

Synonym(s):

Palladium, [2′-[(1,1-dimethylethyl)phenylphosphino-κP]-N2,N2,N6,N6-tetramethyl[1,1′-biphenyl]-2,6-diamine](methanesulfonato-κO)[2′-(methylamino-κN)[1,1′-biphenyl]-2-yl-κC] ChemBeads

Sign Into View Organizational & Contract Pricing

Select a Size

937061-1G
$199.00

$199.00


Please contact Customer Service for Availability


Select a Size

Change View
937061-1G
$199.00

About This Item

Empirical Formula (Hill Notation):
C40H48N3O3PPdS
Molecular Weight:
788.29
UNSPSC Code:
12352100

$199.00


Please contact Customer Service for Availability

form

beads

Quality Level

composition

, 4-6 wt. % (loading)

reaction suitability

reagent type: catalyst

SMILES string

CC(P(C(C=CC=C1)=C1C(C(N(C)C)=CC=C2)=C2N(C)C)C3=CC=CC=C3)(C)C.[MsO][Pd]4C5=CC=CC=C5C6=CC=CC=C6N4C

InChI

InChI=1S/C26H33N2P.C13H12N.CH4O3S.Pd/c1-26(2,3)29(20-14-9-8-10-15-20)24-19-12-11-16-21(24)25-22(27(4)5)17-13-18-23(25)28(6)7;1-14-13-10-6-5-9-12(13)11-7-3-2-4-8-11;1-5(2,3)4;/h8-19H,1-7H3;2-7,9-10,14H,1H3;1H3,(H,2,3,4);/q;-1;;+2/p-1

InChI key

FPQCIPMUSOPSSB-UHFFFAOYSA-M

General description

The ChemBeads product of the (t-Bu)PhCPhos ligand combined with the Buchwald Fourth Generation Palladacycle. It is air, moisture, and thermally-stable and shows good solubility in common organic solvents. Loaded at 5% wt. on glass beads for use in high-throughput expermentation (HTE).

Application

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

For general uses, product is also available in powdered form 900533

Features and Benefits

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryan T Ingoglia et al.
Tetrahedron, 75(32), 4199-4211 (2020-01-04)
Over the past three decades, Pd-catalyzed cross-coupling reactions have become a mainstay of organic synthesis. In particular, catalysts derived from biaryl monophosphines have shown wide utility in forming C-N bonds under mild reaction conditions. This work summarizes a variety of
Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of
Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service