Skip to Content
MilliporeSigma
All Photos(2)

Documents

93524

Supelco

4-(1-Methylhydrazino)-7-nitrobenzofurazan

for HPLC derivatization, LiChropur, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7N5O3
CAS Number:
Molecular Weight:
209.16
Beilstein:
8316412
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for HPLC derivatization

Quality Level

Assay

≥97.0% (HPLC)
≥97.0%

form

powder

quality

LiChropur

technique(s)

HPLC: suitable

mp

~160 °C

storage temp.

2-8°C

SMILES string

CN(N)c1ccc([N+]([O-])=O)c2nonc12

InChI

1S/C7H7N5O3/c1-11(8)4-2-3-5(12(13)14)7-6(4)9-15-10-7/h2-3H,8H2,1H3

InChI key

GVUXVETWVIWDEV-UHFFFAOYSA-N

Other Notes

Superior reagent for monitoring aldehydes and ketones in air; Fluorogenic peroxidase substrate

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Büldt et al.
Analytical chemistry, 71(9), 1893-1898 (1999-05-01)
The synthesis of N-methyl-4-hydrazino-7-nitrobenzofurazan (MNBDH) and its application as a new reagent for the determination of aldehydes and ketones are described. MNBDH reacts with carbonyl compounds in acidic media to the corresponding MNBD-hydrazones. In contrast to the established reagent 2,4-dinitrophenylhydrazine
4-(N-Methylhydrazino)-7-nitro-2,1,3-benzooxadiazole (MNBDH): A Novel Fluorogenic Peroxidase Substrate The authors would like to thank the Fonds der Chemischen Industrie (Frankfurt/Main) for financial support.
Meyer et al.
Angewandte Chemie (International ed. in English), 39(8), 1453-1455 (2000-04-25)
A Büldt et al.
Analytical chemistry, 71(15), 3003-3007 (1999-08-18)
A selective and versatile fluorescence spectroscopic method for the determination of nitrite in waters has been developed. Nitrite reacts in the presence of mineral acids with the nonfluorescent N-methyl-4-hydrazino-7-nitrobenzofurazan forming N-methyl-4-amino-7-nitrobenzofurazan, which can be detected by fluorescence spectroscopy with an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service