Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

92614

Supelco

4-Nitro-7-piperazinobenzofurazan

for HPLC derivatization, LiChropur, ≥99.0%

Synonym(s):

4-Nitro-7-(1-piperazinyl)-2,1,3-benzoxadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5O3
CAS Number:
Molecular Weight:
249.23
Beilstein:
7936339
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for HPLC derivatization

Quality Level

Assay

≥99.0% (HPLC)
≥99.0%

form

powder

quality

LiChropur

technique(s)

HPLC: suitable

fluorescence

λex 395 nm; λem 561 nm in acetone

SMILES string

[O-][N+](=O)c1ccc(N2CCNCC2)c3nonc13

InChI

1S/C10H11N5O3/c16-15(17)8-2-1-7(9-10(8)13-18-12-9)14-5-3-11-4-6-14/h1-2,11H,3-6H2

InChI key

MVLWYDGJBGPXOL-UHFFFAOYSA-N

Application

suitable as derivatizing reagent for isocyanate

Other Notes

Derivatizing agent for the determination of mono- & diisocyanates by LC and MS, UV or fluorescent detection

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hartmut Henneken et al.
Journal of environmental monitoring : JEM, 5(1), 100-105 (2003-03-07)
A diffusive sampling method for the determination of methyl isocyanate (MIC) in air is introduced. MIC is collected using a glass fiber filter impregnated with 4-nitro-7-piperazinobenzo-2-oxa-1,3-diazole (NBDPZ). The urea derivative formed is desorbed from the filter with acetonitrile and analyzed
Chia-Yu Lin et al.
PloS one, 15(10), e0241053-e0241053 (2020-10-27)
The relationship between methylglyoxal (MGO) and D-lactate during saikosaponin C (SSC) treatment of mice with accelerated nephrotoxic serum (NTS) nephritis was investigated. NTS nephritis was induced by administration of anti-basement membrane antibodies to C57BL/6 mice and three dosages of SSC
Shih-Ming Chen et al.
PloS one, 15(7), e0235849-e0235849 (2020-07-11)
Nephrotoxicity severely limits the chemotherapeutic efficacy of cisplatin (CDDP). Oxidative stress is associated with CDDP-induced acute kidney injury (AKI). Methylglyoxal (MG) forms advanced glycation end products that elevate oxidative stress. We aimed to explore the role of MG and its
Yu-Shen Huang et al.
Scientific reports, 8(1), 14587-14587 (2018-10-03)
Chronic kidney disease is a worldwide problem, and Pb contamination is a potential risk factor. Since current biomarkers are not sensitive for the diagnosis of Pb-induced nephrotoxicity, novel biomarkers are needed. Metformin has both hypoglycaemic effects and reno-protection ability. However
Martin Vogel et al.
Analytical chemistry, 74(24), 6418-6426 (2003-01-04)
4-Nitro-7-piperazino-2,1,3-benzoxadiazole (NBDPZ) is presented as a new reagent for the determination of mono- and diisocyanates in air samples. NBDPZ readily reacts with the airborne analytes, thus yielding the corresponding urea derivatives, which are subsequently separated by means of reversed-phase liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service