MilliporeSigma
  • Metabolic profiling of metformin treatment for low-level Pb-induced nephrotoxicity in rat urine.
Metabolic profiling of metformin treatment for low-level Pb-induced nephrotoxicity in rat urine.
Scientific reports (2018-10-03)
Yu-Shen Huang, Shwu-Huey Wang, Shih-Ming Chen, Jen-Ai Lee
ABSTRACT
Chronic kidney disease is a worldwide problem, and Pb contamination is a potential risk factor. Since current biomarkers are not sensitive for the diagnosis of Pb-induced nephrotoxicity, novel biomarkers are needed. Metformin has both hypoglycaemic effects and reno-protection ability. However, its mechanism of action is unknown. We aimed to discover the early biomarkers for the diagnosis of low-level Pb-induced nephrotoxicity and understand the mechanism of reno-protection of metformin. Male Wistar rats were randomly divided into control, Pb, Pb + ML, Pb + MH and MH groups. Pb (250 ppm) was given daily via drinking water. Metformin (50 or 100 mg/kg/d) was orally administered. Urine was analysed by nuclear magnetic resonance (NMR)-based metabolomics coupled with multivariate statistical analysis, and potential biomarkers were subsequently quantified. The results showed that Pb-induced nephrotoxicity was closely correlated with the elevation of 5-aminolevulinic acid, D-lactate and guanidinoacetic acid in urine. After co-treatment with metformin, 5-aminolevulinic acid and D-lactate were decreased. This is the first demonstration that urinary 5-aminolevulinic acid, D-lactate and guanidinoacetic acid could be early biomarkers of low-level Pb-induced nephrotoxicity in rats. The reno-protection of metformin might be attributable to the reduction of D-lactate excretion.
Product Description
Supelco
3-(Trimethylsilyl)propionic-2,2,3,3-d4 acid sodium salt, 98 atom % D
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Guanidineacetic acid, 99%
Sigma-Aldrich
Indole-2-acetic acid