Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92358

Supelco

Thioanisole

analytical standard

Synonym(s):

Methyl phenyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SCH3
CAS Number:
Molecular Weight:
124.20
Beilstein:
1904179
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.586-1.588
n20/D 1.587 (lit.)

bp

188 °C (lit.)

mp

−15 °C (lit.)

density

1.057 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CSc1ccccc1

InChI

1S/C7H8S/c1-8-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

HNKJADCVZUBCPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy H Van Hove et al.
Biomaterials, 35(36), 9719-9730 (2014-09-03)
Despite the recent expansion of peptide drugs, delivery remains a challenge due to poor localization and rapid clearance. Therefore, a hydrogel-based platform technology was developed to control and sustain peptide drug release via matrix metalloproteinase (MMP) activity. Specifically, hydrogels were
Yunfei Sun et al.
International journal of pharmaceutics, 468(1-2), 83-90 (2014-04-09)
Gene therapy is a potent tool for the treatment of cancer and other gene defect diseases, which involves using DNA that encodes a functional, therapeutic gene to replace a mutated gene. However, the DNA transfection efficiency is restricted by its
B Frohm et al.
Soft matter, 11(2), 414-421 (2014-11-20)
The peptide GSFSIQYTYHV derived from human semenogelin I forms a transparent hydrogel through spontaneous self-assembly in water at neutral pH. Linear rheology measurements demonstrate that the gel shows a dominating elastic response over a large frequency interval. CD, fluorescence and
Veronika Mäde et al.
ChemMedChem, 9(11), 2463-2474 (2014-08-27)
Pancreatic polypeptide (PP) is a satiety-inducing gut hormone targeting predominantly the Y4 receptor within the neuropeptide Y multiligand/multireceptor family. Palmitoylated PP-based ligands have already been reported to exert prolonged satiety-inducing effects in animal models. Here, we suggest that other lipidation
Lisa Pollaro et al.
Molecular cancer therapeutics, 14(1), 151-161 (2014-11-09)
Monoclonal antibodies have long in vivo half-lives and reach high concentrations in tumors but cannot access all regions in the tissue, whereas smaller ligands such as peptides distribute better but are limited by low concentrations due to fast renal clearance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service