Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90867

Supelco

(−)-Methyl L-lactate

analytical standard

Synonym(s):

Methyl (S)-(−)-lactate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CO2CH3
CAS Number:
Molecular Weight:
104.10
Beilstein:
1720587
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

Quality Level

grade

analytical standard

Assay

≥98.5% (GC)

optical purity

enantiomeric excess: ≥97.0%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

refractive index

n20/D 1.413 (lit.)
n20/D 1.413-1.415

bp

144-145 °C (lit.)

density

1.09 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COC(=O)[C@H](C)O

InChI

1S/C4H8O3/c1-3(5)4(6)7-2/h3,5H,1-2H3/t3-/m0/s1

InChI key

LPEKGGXMPWTOCB-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service