Skip to Content
MilliporeSigma
(−)-Methyl L-lactate 98%, optical purity ee: 97% (GLC)

(−)-Methyl L-lactate

(−)-Methyl L-lactate 98%, optical purity ee: 97% (GLC)
Synonym(s):
Methyl (S)-(−)-lactate
Linear Formula:
CH3CH(OH)CO2CH3
CAS Number:
Molecular Weight:
104.10
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing