Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87649

Sigma-Aldrich

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym(s):

(S)-(+)-α-Tetralol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O
CAS Number:
Molecular Weight:
148.20
Beilstein:
2208779
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

grade

puriss.

Assay

≥99.0% (sum of enantiomers, GC)

form

crystals

optical activity

[α]20/D +33±1°, c = 2.5% in chloroform

optical purity

enantiomeric ratio: 99:1 (GC)

mp

37-39 °C

SMILES string

O[C@H]1CCCc2ccccc12

InChI

1S/C10H12O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-2,4,6,10-11H,3,5,7H2/t10-/m0/s1

InChI key

JAAJQSRLGAYGKZ-JTQLQIEISA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urmi Dhagat et al.
Bioorganic & medicinal chemistry letters, 20(17), 5274-5276 (2010-07-27)
A comparison of the structures and kinetic properties of human 20alpha-hydroxysteroid dehydrogenase (AKR1C1) and its mutant enzymes (Leu308Val and Leu308Ala) indicates that Leu308 is a selectivity determinant for substrate binding. While the Leu308Val mutation improved the catalytic efficiency (k(cat)/K(m)) of
Ingo Bichlmaier et al.
Journal of medicinal chemistry, 49(5), 1818-1827 (2006-03-03)
A set of 28 enantiomers comprising rigid and flexible secondary alcohols was synthesized by the asymmetric Corey-Bakshi-Shibata reduction. The enantiomerically pure alcohols were subjected to enzymatic glucuronidation assays employing the human UDP-glucuronosyltransferases (UGTs) 2B7 and 2B17. Both UGTs displayed high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service