Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

80337

Supelco

(±)-Metoprolol (+)-tartrate salt

analytical standard

Synonym(s):

(±)1-(Isopropylamino)-3-[p-(β-methoxyethyl)phenoxy]-2-propanol (+)-tartrate salt, Lopressor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C15H25NO3)2 · C4H6O6
CAS Number:
Molecular Weight:
684.81
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.COCCc1ccc(OCC(O)CNC(C)C)cc1.COCCc2ccc(OCC(O)CNC(C)C)cc2

InChI

1S/2C15H25NO3.C4H6O6/c2*1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3;5-1(3(7)8)2(6)4(9)10/h2*4-7,12,14,16-17H,8-11H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1

InChI key

YGULWPYYGQCFMP-CEAXSRTFSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V M Raju et al.
Clinical radiology, 69(7), 739-744 (2014-05-16)
To evaluate the usage, safety, and efficacy of high-dose intravenous metoprolol for heart rate reduction in computer tomographic (CT) coronary angiography. As this was retrospective analysis of anonymous data, medical ethics committee approval was waived by the regional health research
Wenhuan Li et al.
Medicine, 93(28), e329-e329 (2014-12-20)
The sensitivity and specificity of 5 different image sets of dual-energy computed tomography (DECT) for the detection of first-pass myocardial perfusion defects have not systematically been compared using positron emission tomography (PET) as a reference standard. Forty-nine consecutive patients, with
Rahil H Kassamali et al.
AJR. American journal of roentgenology, 203(4), 759-762 (2014-09-24)
The purpose of this study was to assess the safety of heart rate optimization by use of β-adrenergic blockade solely by the i.v. route before coronary CT angiography. The records of 679 patients undergoing CT coronary angiography after receiving i.v.
I S Hamadeh et al.
Clinical pharmacology and therapeutics, 96(2), 175-181 (2014-03-19)
Metoprolol is a selective β-1 adrenergic receptor blocker that undergoes extensive metabolism by the polymorphic enzyme cytochrome P450 2D6 (CYP2D6). Our objective was to investigate the influence of CYP2D6 polymorphisms on the efficacy and tolerability of metoprolol tartrate. Two hundred
Reply: Carvedilol versus metoprolol, but which metoprolol? Effect on inappropriate cardioverter-defibrillator therapy.
Martin H Ruwald et al.
Journal of the American College of Cardiology, 63(18), 1933-1934 (2014-02-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service