Skip to Content
MilliporeSigma
(±)-Metoprolol (+)-tartrate salt ≥98% (titration), powder

(±)-Metoprolol (+)-tartrate salt

(±)-Metoprolol (+)-tartrate salt ≥98% (titration), powder
Synonym(s):
(±)1-(Isopropylamino)-3-[p-(β-methoxyethyl)phenoxy]-2-propanol (+)-tartrate salt, Lopressor
Linear Formula:
(C15H25NO3)2 · C4H6O6
CAS Number:
Molecular Weight:
684.81
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing