Skip to Content
MilliporeSigma
All Photos(1)

Documents

76721

Millipore

XLT4 Agar (Base)

NutriSelect® Plus, suitable for microbiology

Synonym(s):

Xylose Lysine Tergitol-4 Agar (Base)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 18 g/L
ammonium-iron(III) citrate, 0.8 g/L
lactose, 7.5 g/L
L-lysine, 5 g/L
phenol-red, 0.08 g/L
proteose peptone No. 3, 1.6 g/L
sodium chloride, 5 g/L
sodium thiosulfate, 6.8 g/L
sucrose, 7.5 g/L
xylose, 3.75 g/L
yeast extract, 3 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages

microbiology

suitability

selective and differential for Citrobacter spp.
selective and differential for Escherichia coli
selective and differential for Salmonella spp.
selective and differential for Shigella spp.
selective and differential for coliforms
selective and differential for enterobacteriaceae

Related Categories

Application

for the isolation and identification of pathogenic Enterobacteriaceae, especially Salmonella species

Preparation Note

Suspend 59 g in 1 litre distilled water, add 4.6 ml XLT4 Agar Supplement solution (Cat. No. 83714) and heat the medium in a boiling water-batch (not on a heating-plate!). Cool to approx. 50°C and pour plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service