Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.08981

Millipore

XLT-4 Agar Supplement

for Salmonella spp., pkg of 100 mL, for use with GranuCult® XLT4 agar (Base)

Synonym(s):

Salmonella Selective Supplement

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$77.60

$77.60


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
100 ML
$77.60

About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

$77.60


Please contact Customer Service for Availability

Request more information

form

liquid

Quality Level

composition

Sodium tetradecylsulfate solution, 26-28%

packaging

pkg of 100 mL

technique(s)

microbiological culture: suitable

bp

100 °C

density

1.06 g/cm3

application(s)

food and beverages
microbiology

compatibility

for use with GranuCult® XLT4 agar (Base)

storage temp.

15-25°C

suitability

Salmonella spp.
enterobacteriaceae

General description

XLT-4 Agar Supplement contains sodiumtetradecylsulfate solution (26-28 %). It is a surfactant that inhibits thegrowth of accompanying flora and is used during the preparation of XLT-4 AgarBase to isolate and identify pathogenic Enterobacteriaceae.

Application

XLT-4 Agar Supplement is an additive used during the preparation of XLT-4 Agar Base for the isolation and identification of pathogenic Enterobacteriaceae, especially Salmonella spp.

Analysis Note

Growth
- Salmonella enteritidis ATCC 13076 (WDCM 00030):
good to very good
- Salmonella typhimurium ATCC 14028 (WDCM 00031): good to very good
- Salmonella anatum ATCC 9270: fair to very good
- Shigella sonnei ATCC 11060: fair to very good
- Shigella flexneri ATCC 12022 (WDCM 00126): good to very good
- Enterobacter aerogenes ATCC 13048: fair to good
- Citrobacter freundii ATCC 8090: fair to good
- Escherichia coli ATCC 25922 (WDCM 00013): none to fair
- Proteus mirabilis ATCC 14273: none to poor
Black centre
- Salmonella enteritidis ATCC 13076 (WDCM 00030):
+
- Salmonella typhimurium ATCC 14028 (WDCM 00031): +
- Salmonella anatum ATCC 9270: +
Colony colour
- Shigella sonnei ATCC 11060:
colourless
- Shigella flexneri ATCC 12022 (WDCM 00126): colourless
- Enterobacter aerogenes ATCC 13048: yellow
- Citrobacter freundii ATCC 8090: yellow
- Escherichia coli ATCC 25922 (WDCM 00013): colourless to yellow
Incubation: 24 - 48 hrs.; 35 °C.
Supplement tested with XLT4 agar base Art. 1.08981.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Belgin Siriken et al.
Microbial drug resistance (Larchmont, N.Y.), 26(2), 136-144 (2019-08-28)
The aims of this study were to evaluate the prevalence of Salmonella spp., including S. Enteritidis and S. Typhimurium, their antibiotic resistance profiles, and the presence/absence of class 1 integron (intI1) in 50 raw ground beef and 50 raw, meatball

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service