Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

74196

Supelco

Zinc ionophore IV

Selectophore, function tested

Synonym(s):

Potassium hydrotris(N-tert-butyl-2-thioimidazolyl)borate, [TmBut]K

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34BKN6S3
CAS Number:
Molecular Weight:
516.64
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

powder

quality

function tested

SMILES string

[K+].CC(C)(C)N1C=CN([BH-](N2C=CN(C2=S)C(C)(C)C)N3C=CN(C3=S)C(C)(C)C)C1=S

InChI

1S/C21H34BN6S3.K/c1-19(2,3)23-10-13-26(16(23)29)22(27-14-11-24(17(27)30)20(4,5)6)28-15-12-25(18(28)31)21(7,8)9;/h10-15,22H,1-9H3;/q-1;+1

InChI key

ZYQDTWROYIMHBC-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Application

Used for selective determination of Zn(II) in biological and environmental samples.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashok Kumar Singh et al.
Analytical and bioanalytical chemistry, 388(8), 1867-1876 (2007-07-12)
Potassium hydrotris(N-tert-butyl-2-thioimidazolyl)borate [KTtt-Bu] and potassium hydrotris(3-tert-butyl-5-isopropyl-l-pyrazolyl)borate [KTpt-Bu,i-Pr] have been synthesized and evaluated as ionophores for preparation of a poly(vinyl chloride) (PVC) membrane sensor for Zn(II) ions. The effect of different plasticizers, viz. benzyl acetate (BA), dioctyl phthalate (DOP), dibutyl phthalate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service