Skip to Content
MilliporeSigma
All Photos(1)

Documents

73741

Supelco

2-Nitrophenyl pentyl ether

Selectophore, ≥99.0%

Synonym(s):

1-Nitro-2-pentyloxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO3
CAS Number:
Molecular Weight:
209.24
Beilstein:
2105866
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥99.0% (GC)
≥99.0%

form

liquid

refractive index

n20/D 1.522

bp

177-178 °C/15 mmHg (lit.)

density

1.098 g/mL at 20 °C (lit.)

SMILES string

CCCCCOc1ccccc1[N+]([O-])=O

InChI

1S/C11H15NO3/c1-2-3-6-9-15-11-8-5-4-7-10(11)12(13)14/h4-5,7-8H,2-3,6,9H2,1H3

InChI key

YRVCISCRAMHCFP-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Plasticizer used in polymer membrane neutral carrier-based ion-selective electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Drouin et al.
Metabolites, 10(1) (2019-12-22)
Cardiovascular diseases (CVDs) represent a major concern in today's society, with more than 17.5 million deaths reported annually worldwide. Recently, five metabolites related to the gut metabolism of phospholipids were identified as promising predictive biomarker candidates for CVD. Validation of
A S Attiyat et al.
Talanta, 35(10), 789-794 (1988-10-01)
Lithium, potassium and caesium-selective microelectrodes were prepared by coating the tips of preconditioned silver wires, incorporated in a flow-cell, with PVC membranes containing four different ionophores. A dicarboxamide, a 14-crown-4 carboxylic acid, benzo-18-crown-6 and di-(tert-butylbenzo)-21-crown-7 ionophores were used in the
A.S. Attiyat et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 37, 122-122 (1988)
Magnus Saed Restan et al.
Analytica chimica acta, 1104, 1-9 (2020-02-29)
Electromembrane extraction (EME) in small, stagnant and chip-like devices has the potential for future in-field operation. Literature briefly discuss such systems, but performance suffered from evaporative losses of sample and acceptor. To address this, the current paper reports electromembrane extraction
Elzbieta Radzyminska-Lenarcik et al.
Membranes, 10(5) (2020-05-06)
Polymer inclusion membranes (PIMs) doped with ethylenodiamino-bis-acetylacetone as fixed carrier was applied for the investigation of the facilitated transport of Zn(II), Cr(III), and Ni(II) ions from an aqueous nitrate feed phase (cM = 0.001 mol/dm3). The optimal membrane composition (amount

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service