MilliporeSigma
  • A comparative study of the effect of o-nitrophenyl octyl ether and o-nitrophenyl pentyl ether as plasticizers on the response and selectivity of carrier-based liquid membrane ion-selective electrodes.
A comparative study of the effect of o-nitrophenyl octyl ether and o-nitrophenyl pentyl ether as plasticizers on the response and selectivity of carrier-based liquid membrane ion-selective electrodes.
Talanta (1988-10-01)
A S Attiyat, G D Christian, J L Hallman, R A Bartsch
ABSTRACT
Lithium, potassium and caesium-selective microelectrodes were prepared by coating the tips of preconditioned silver wires, incorporated in a flow-cell, with PVC membranes containing four different ionophores. A dicarboxamide, a 14-crown-4 carboxylic acid, benzo-18-crown-6 and di-(tert-butylbenzo)-21-crown-7 ionophores were used in the electrode matrix. The first two ionophores were used in lithium ion-selective electrodes, the third in a potassium ion electrode and the fourth in a caesium ion electrode. Two different plasticizers, o-nitrophenyl octyl ether (NPOE) and o-nitrophenyl pentyl ether (NPP'E) were used. Enhancement of the signal and the slope of the calibration curve and improvement of the curve linearity were observed in all cases when NPP'E was used as plasticizer. A general trend of enhanced selectivity of the electrodes incorporating crown ether ionophores was also observed when NPP'E was the plasticizer.