Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

67988

Supelco

Punicalin, mixture of anomers

analytical standard

Synonym(s):

4,6-Gallagyl-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H22O22
CAS Number:
Molecular Weight:
782.53
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

OC1O[C@@H]2COC(=O)c3cc(O)c(O)c(O)c3-c4c(O)c(O)c5OC(=O)c6c(c(O)c(O)c7OC(=O)c4c5-c67)-c8c(O)c(O)c(O)cc8C(=O)O[C@H]2[C@H](O)[C@H]1O

InChI

1S/C34H22O22/c35-6-1-4-9(19(39)17(6)37)11-15-13-14-16(33(50)56-28(13)23(43)21(11)41)12(22(42)24(44)29(14)55-32(15)49)10-5(2-7(36)18(38)20(10)40)31(48)54-27-8(3-52-30(4)47)53-34(51)26(46)25(27)45/h1-2,8,25-27,34-46,51H,3H2/t8-,25-,26-,27-,34-/m1/s1

InChI key

IQHIEHIKNWLKFB-ITTSEVFZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C C Lin et al.
Anticancer research, 21(1A), 237-243 (2001-04-13)
Reactive oxygen species (ROS) react with biological molecules and destroy the structure of cells and eventually cause free radical-induced disease such as inflammation and cancer. Previous studies showed that an aqueus extract of Terminalia catappa L. exhibited superoxide radical scavenger
C C Lin et al.
The American journal of Chinese medicine, 27(3-4), 371-376 (1999-12-11)
Punicalagin and punicalin were isolated from the leaves of Terminalia catappa L. In this study, we evaluated the anti-inflammatory activity of punicalagin and punicalin carrageenan-induced hind paw edema in rats. After evaluation of the anti-inflammatory effects, the edema rates were
V Martino et al.
Phytotherapy research : PTR, 18(8), 667-669 (2004-10-09)
The bioassay- guided fractionation of the aqueous extract of Terminalia triflora leaves afforded punicalin and 2-O-galloylpunicalin, isolated for the first time from this species. These compounds showed inhibitory activity on HIV-1 reverse transcriptase in a dose-dependent manner. Punicalin showed an
C C Lin et al.
The Journal of pharmacy and pharmacology, 50(7), 789-794 (1998-08-28)
Punicalagin and punicalin, isolated from the leaves of Terminalia catappa L., are used to treat dermatitis and hepatitis. Both compounds have strong antioxidative activity. The antihepatotoxic activity of punicalagin and punicalin on carbon tetrachloride (CCl4)-induced toxicity in the rat liver
M Jimenez Del Rio et al.
Georgian medical news, (140)(140), 70-77 (2006-12-21)
This study analyzes the major phenolic constituents of pomegranate fruit juice and pericarp feedstock, and dry extracts thereof, using high-performance liquid chromatography. Pomegranate pericarp was extracted with water (WE) and alcohol (AE) as solvents, and liquid extracts were subsequently freeze-dried.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service