Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHL80524

Punicalagin (A + B mixture)

phyproof® Reference Substance

Synonym(s):

Punicalagin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H28O30
CAS Number:
Molecular Weight:
1084.72
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

OC1O[C@@H]2COC(=O)c3cc(O)c(O)c(O)c3-c4c(O)c(O)c5OC(=O)c6c(c(O)c(O)c7OC(=O)c4c5-c67)-c8c(O)c(O)c(O)cc8C(=O)O[C@H]2[C@@H]9OC(=O)c%10cc(O)c(O)c(O)c%10-c%11c(O)c(O)c(O)cc%11C(=O)O[C@@H]19

InChI

1S/C48H28O30/c49-10-1-6-17(31(59)27(10)55)19-23-21-22-24(47(70)76-38(21)35(63)33(19)61)20(34(62)36(64)39(22)75-46(23)69)18-9(4-13(52)28(56)32(18)60)43(66)74-37-14(5-72-42(6)65)73-48(71)41-40(37)77-44(67)7-2-11(50)25(53)29(57)15(7)16-8(45(68)78-41)3-12(51)26(54)30(16)58/h1-4,14,37,40-41,48-64,71H,5H2/t14-,37-,40+,41-,48?/m1/s1

InChI key

ZJVUMAFASBFUBG-UYMKNUMKSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Punicalagin as a Bioactive Compound in Biotech Research: Punicalagin demonstrates significant potential in reducing hyperuricemia-related renal and intestinal dysfunctions, highlighting its role in pharmaceutical research focused on anti-inflammatory and antioxidant therapies (Han et al., 2024).

  • Antioxidant Properties of Punicalagin in Biochemistry: Investigating the antioxidant action of pomegranate extract containing punicalagin on eroded dentin reveals insights into its mechanisms of action at the molecular level, which is crucial for developing advanced dental therapeutic materials (Cortez et al., 2024).

  • Punicalagin A and B Mixture for Laboratory Research: Studies on the effects of punicalagin on stress urinary incontinence demonstrate the compound′s therapeutic potential via the upregulation of Nrf2 and TGF-β1 signaling pathways, supporting its use in biotechnological applications related to urogenital health (Tang et al., 2024).

Biochem/physiol Actions

Potent immune suppressant isolated from pomegranate.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service