Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

66450

Sigma-Aldrich

4-Methyl-1-cyclohexene

purum, ≥99.0% (GC)

Synonym(s):

1,2,3,6-Tetrahydrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12
CAS Number:
Molecular Weight:
96.17
Beilstein:
1901299
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥99.0% (GC)

refractive index

n20/D 1.442

bp

101-102 °C (lit.)

density

0.799 g/mL at 20 °C (lit.)

SMILES string

CC1CCC=CC1

InChI

1S/C7H12/c1-7-5-3-2-4-6-7/h2-3,7H,4-6H2,1H3

InChI key

FSWCCQWDVGZMRD-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service