Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.51056

Sigma-Aldrich

HOSu

Novabiochem®

Synonym(s):

HOSu, 1-Hydroxy-2,5-pyrrolidinedione, N-Hydroxysuccinimide, NHS

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$31.10
100 G
$106.00
1 KG
$450.00

$31.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$31.10
100 G
$106.00
1 KG
$450.00

About This Item

Empirical Formula (Hill Notation):
C4H5NO3
CAS Number:
Molecular Weight:
115.09
MDL number:
UNSPSC Code:
12352005
EC Index Number:
228-001-3
NACRES:
NA.22

$31.10


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥99.0% (acidimetric)

form

powder

reaction suitability

reaction type: Coupling Reactions

manufacturer/tradename

Novabiochem®

pH

5-7 (20 °C, 50 g/L in H2O)

mp

93-100 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N1(C(=O)CCC1=O)O

InChI

1S/C4H5NO3/c6-3-1-2-4(7)5(3)8/h8H,1-2H2

InChI key

NQTADLQHYWFPDB-UHFFFAOYSA-N

General description

Reagent used for the synthesis of water-soluble active esters and as an additive to suppress enantiomerization during carbodiimide condensations [1,2].

Associated Protocols and Technical Articles
Guide to Selection of Coupling Reagents

Literature references

[1] G. W. Anderson, et al. (1964) J. Am. Chem. Soc., 86, 1839.
[2] F. Weygand, et al. (1966) Z. Naturforsch., 21, 426.

Linkage

Replaces: 01-62-0009

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 95 °C
Melting range (upper value): ≤ 100 °C
Solubility (1 mmole in 2 ml DMF): clearly soluble
Identity (IR): passes test

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service