Skip to Content
MilliporeSigma
All Photos(1)

Documents

65209

Supelco

(R)-(−)-α-Methoxyphenylacetic acid

for chiral derivatization, LiChropur, ≥99.0%

Synonym(s):

O-Methyl-D-mandelic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OCH3)CO2H
CAS Number:
Molecular Weight:
166.17
Beilstein:
2209198
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for chiral derivatization

Quality Level

Assay

≥99.0% (T)
≥99.0%

form

solid

optical activity

[α]20/D -150±3°, c = 0.5% in ethanol

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

quality

LiChropur

technique(s)

HPLC: suitable

mp

64-67 °C
66-68 °C (lit.)

SMILES string

CO[C@@H](C(O)=O)c1ccccc1

InChI

1S/C9H10O3/c1-12-8(9(10)11)7-5-3-2-4-6-7/h2-6,8H,1H3,(H,10,11)/t8-/m1/s1

InChI key

DIWVBIXQCNRCFE-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(−)-α-Methoxyphenylacetic acid is a chiral derivatizating agent, which is employed for derivatizing enantiomers into diastereoisomers.

Other Notes

Chiral reagent for the det. of enantiomeric purity and absolute configuration of sec-alcohols and α-chiral amines by NMR; Analysis of amines using the ONSu ester

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Husain et al.
Analytical biochemistry, 178(1), 177-183 (1989-04-01)
A chiral derivatizing reagent, N-succinimidyl-2-(S)-methoxy-2-phenylacetic acid ester (SMPA), directed toward reaction with primary amine-containing compounds has been synthesized and characterized. This reagent is suitable for HPLC resolution from enzymatic-scale reactions where only microgram quantities of chiral products may be obtainable.
The Journal of Organic Chemistry, 59, 4202-4202 (1994)
B.M. Trost et al.
The Journal of Organic Chemistry, 51, 2370-2370 (1986)
P A Husain et al.
Analytical chemistry, 65(10), 1456-1461 (1993-05-15)
We introduce a novel, HPLC-based method for facile determination of the absolute configuration of alpha-chiral amines. Our method is easily applied to a variety of compounds, including amino acid derivatives. The method involves initial derivatization of the chiral amine analyte
Radoslaw Laufer et al.
Bioorganic & medicinal chemistry, 22(17), 4968-4997 (2014-07-22)
TTK kinase was identified by in-house siRNA screen and pursued as a tractable, novel target for cancer treatment. A screening campaign and systematic optimization, supported by computer modeling led to an indazole core with key sulfamoylphenyl and acetamido moieties at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service