MilliporeSigma
  • N-succinimidyl methoxyphenylacetic acid ester, an amine-directed chiral derivatizing reagent suitable for enzymatic scale resolutions.
N-succinimidyl methoxyphenylacetic acid ester, an amine-directed chiral derivatizing reagent suitable for enzymatic scale resolutions.
Analytical biochemistry (1989-04-01)
P A Husain, J E Colbert, S R Sirimanne, D G VanDerveer, H H Herman, S W May
ABSTRACT
A chiral derivatizing reagent, N-succinimidyl-2-(S)-methoxy-2-phenylacetic acid ester (SMPA), directed toward reaction with primary amine-containing compounds has been synthesized and characterized. This reagent is suitable for HPLC resolution from enzymatic-scale reactions where only microgram quantities of chiral products may be obtainable. SMPA derivatization was shown to be effective in the resolution of the enantiomers of a number of different racemic compounds. SMPA was used to resolve the diastereoisomeric derivatives of a previously unknown enzymatically oxygenated product, allowing determination of the stereochemical course of the enzymatic reaction. SMPA is easily prepared from an inexpensive, commercially available, and enantiomerically pure precursor with the formation of a shelf-stable crystalline product which is utilizable in water-containing solutions. In addition to its usefulness for micro-determinations, SMPA is useful for preparative-scale resolutions of enantiomers since the reagent is cleaved from the diastereoisomeric derivative by acid hydrolysis.