Skip to Content
MilliporeSigma
All Photos(1)

Documents

61471

Millipore

ESBL ChromoSelect Agar Supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Streptococcus spp.

Application

Strep B ChromoSelect Selective Agar Base is used for the detection of Extended Spectrum Beta lactamase producing organisms.

Components

Composition:
(per vial sufficient for 500 ml medium)
Ceftazidime 1.50 mg
Cefotaxime 1.50 mg
Ceftriazone 1.00 mg
Aztreonam 1.00 mg
Fluconazole 5.00 mg

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service