Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55286

Millipore

VRE ChromoSelect Agar Supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

5 VIALS
$175.00

$175.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
5 VIALS
$175.00

About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

$175.00


Please contact Customer Service for Availability

Request more information

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Enterococcus spp.

Application

VRE ChromoSelect Agar Supplement is recommended for selective isolation of Vancomycin Resistant Enterococci (VRE).

Components

Composition (per vial, sufficient for 500 ml)
Vancomycin 4mg
Fluconazole 5mg

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service