Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59754

Supelco

Steviolbioside

analytical standard

Synonym(s):

(4α)-13-[(2-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]kaur-16-en-18-oic acid, Steviobioside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H50O13
CAS Number:
Molecular Weight:
642.73
UNSPSC Code:
85151701

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤12.0% water (Karl Fischer)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

C[C@@]12CCC[C@](C)([C@H]1CC[C@]34CC(=C)[C@](CC[C@@H]23)(C4)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C(O)=O

InChI

1S/C32H50O13/c1-15-11-31-9-5-18-29(2,7-4-8-30(18,3)28(40)41)19(31)6-10-32(15,14-31)45-27-25(23(38)21(36)17(13-34)43-27)44-26-24(39)22(37)20(35)16(12-33)42-26/h16-27,33-39H,1,4-14H2,2-3H3,(H,40,41)/t16-,17-,18-,19-,20-,21-,22+,23+,24-,25-,26+,27+,29+,30-,31+,32-/m1/s1

InChI key

OMHUCGDTACNQEX-VKXXIMTPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lun-Huei Lin et al.
Chemical & pharmaceutical bulletin, 52(9), 1117-1122 (2004-09-02)
A new group of steviolbioside amide dimers 2a-g, derivatives 2h-i and their related steviol and isosteviol amide dimers 3a and 4a were prepared by reacting aliphatic alkylamine and alkyldiamines with PyBOP and DIEA. The synthesized compounds had cytotoxic effects on
Vikas Jaitak et al.
Journal of pharmaceutical and biomedical analysis, 47(4-5), 790-794 (2008-05-06)
A high-performance thin-layer chromatographic (HPTLC) method was developed and validated as per ICH (International Conferences on Harmonization) guidelines for simultaneous quantification of three steviol glycosides, i.e. steviolbioside, stevioside and rebaudioside-A in Stevia rebaudiana leaves. For achieving good separation, mobile phase
K Ohtani et al.
Chemical & pharmaceutical bulletin, 39(12), 3172-3174 (1991-12-01)
1,4-alpha-Glucosylation at the 13-O-glycosyl moiety of stevioside (S) and rubusoside (RU) results in a significant increase of sweetness. Saponification of the 19-COO-beta-glucosyl linkage of S and RU yielded steviolbioside (SB) (= 13-O-beta-sophorosyl-steviol) and steviolmonoside (SM) (= 13-O-beta-glucosyl-steviol), respectively, both of
V E Kataev et al.
Bioorganicheskaia khimiia, 37(4), 542-551 (2011-11-22)
Conjugates of antitubercular drug Isoniazid (hydrazide of isonicotinic acid), nicotinic and alpha-picolinic acid hydrazides and glycoside steviolbioside from the plant Stevia rebaudiana as well as the product of its acid hydrolysis, diterpenoid isosteviol, were synthesized. Besides, isosteviol hydrazide and hydrazone
Karolina Jaworska et al.
Journal of AOAC International, 95(6), 1588-1596 (2013-03-05)
Simultaneous separation of steviol and steviol glycosides is challenging because of differences in their polarity and chemical structure. In this study, simultaneous analysis of steviol and steviol glycosides was achieved by LC with UV detection using a mixed-mode RP weak

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service