MilliporeSigma
  • Validated high-performance thin-layer chromatography method for steviol glycosides in Stevia rebaudiana.
Validated high-performance thin-layer chromatography method for steviol glycosides in Stevia rebaudiana.
Journal of pharmaceutical and biomedical analysis (2008-05-06)
Vikas Jaitak, A P Gupta, V K Kaul, P S Ahuja
ABSTRACT
A high-performance thin-layer chromatographic (HPTLC) method was developed and validated as per ICH (International Conferences on Harmonization) guidelines for simultaneous quantification of three steviol glycosides, i.e. steviolbioside, stevioside and rebaudioside-A in Stevia rebaudiana leaves. For achieving good separation, mobile phase of ethyl acetate-ethanol-water (80:20:12, v/v/v) on pre-coated silica gel 60 F254 HPTLC plates were used. The densitometric quantification of steviol glycosides was carried out at lambda=510 nm in reflection-absorption mode after spraying with acetic anhydride:sulphuric acid:ethanol reagent. The calibration curves were linear in the range of 160-960 ng/spot for steviolbioside, 1-6 microg/spot for stevioside and 0.5-3 microg/spot for rebaudioside-A with good correlation coefficients (0.998-0.999). The method was found to be reproducible for quantitative analysis of steviol glycosides in S. rebaudiana leaves collected from ten different locations and will serve as a quality control indicator to monitor the commercial production of stevioside and its allied molecules during different stages of its processing.
Sigma-Aldrich
Steviol hydrate, ≥98% (HPLC), powder