Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57153

Supelco

ddNTP Set

suitable for MALDI-MS

Synonym(s):

Dideoxynucleoside Triphosphate Set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

composition

ddATP, 10 mmol/L
ddCTP, 10 mmol/L
ddGTP, 10 mmol/L
ddTTP, 10 mmol/L

technique(s)

MALDI-MS: suitable

storage temp.

−20°C

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y H Liu et al.
Rapid communications in mass spectrometry : RCM, 9(9), 735-743 (1995-01-01)
A new approach is developed for the rapid and cost-effective detection of human genetic polymorphisms based on matrix-assisted laser description/ionization mass spectrometric (MALDI MS) detection using a nitrocellulose film substrate. This method employs polymerase chain reaction (PCR) amplification using DNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service