Skip to Content
MilliporeSigma
All Photos(1)

Documents

54300

Sigma-Aldrich

Silica gel Rubin drying bags

with indicator function, 4 TE, bag with ~120 g, crystalline

Synonym(s):

Drying bags with silicagel Rubin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101527

form

crystalline

quality

4 TE
bag with ~120 g
with indicator function

manufacturer/tradename

Zeochem AG

particle size

1.0-3.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

active: red; inactive: yellow; regeneration at max. 80°C

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service