Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

53527

Sigma-Aldrich

L-Homocysteine thiolactone hydrochloride

≥99.0% (TLC)

Synonym(s):

L-2-Amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NOS · HCl
CAS Number:
Molecular Weight:
153.63
Beilstein:
3911455
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99.0% (TLC)

form

powder

optical activity

[α]20/D +22.5±1°, c = 2.5% in 5 M HCl

technique(s)

LC/MS: suitable

color

white

mp

185-191 °C

application(s)

peptide synthesis

SMILES string

Cl[H].N[C@H]1CCSC1=O

InChI

1S/C4H7NOS.ClH/c5-3-1-2-7-4(3)6;/h3H,1-2,5H2;1H/t3-;/m0./s1

InChI key

ZSEGSUBKDDEALH-DFWYDOINSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service