Skip to Content
MilliporeSigma
L-Homocysteine thiolactone hydrochloride ≥98% (TLC)

L-Homocysteine thiolactone hydrochloride

L-Homocysteine thiolactone hydrochloride ≥98% (TLC)
Synonym(s):
L-2-Amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride
Empirical Formula (Hill Notation):
C4H7NOS · HCl
CAS Number:
Molecular Weight:
153.63
EC Number:

Products

Product Category
Color
Formula Weight
Physical Form
Purity
Application
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing