Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51773

Supelco

Desmethoxyyangonin

analytical standard

Synonym(s):

(E)-4-Methoxy-6-(2-phenylethenyl)-2H-pyran-2-one, 5,6-Dehydrokavain, Demethoxyyangonin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24
Beilstein:
181238
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COC1=CC(=O)OC(\C=C\c2ccccc2)=C1

InChI

1S/C14H12O3/c1-16-13-9-12(17-14(15)10-13)8-7-11-5-3-2-4-6-11/h2-10H,1H3/b8-7+

InChI key

DKKJNZYHGRUXBS-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atul Upadhyay et al.
Journal of agricultural and food chemistry, 59(7), 2857-2862 (2011-02-11)
AIDS and influenza are viral pandemics and remain one of the leading causes of human deaths worldwide. The increasing resistance of these diseases to synthetic drugs demands the search for novel compounds from plant-based sources. In this regard, the leaves
[Effects of the constituents of Alpinia speciosa rhizoma on experimental ulcers].
S Y Hsu
Taiwan yi xue hui za zhi. Journal of the Formosan Medical Association, 86(1), 58-64 (1987-01-01)
Yen Thi-Kim Nguyen et al.
Molecules (Basel, Switzerland), 24(12) (2019-06-21)
Pavetta indica L. is used in traditional medicine for the treatment of various diseases including hemorrhoids, headache, urinary conditions, ulcerated nose, and dropsy. However, no study has evaluated the anticancer effect of P. indica L. In this study, we found
I Jantan et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 15(4), 306-309 (2007-10-05)
Twelve compounds isolated from Alpinia mutica Roxb., Kaempferia rotunda Linn., Curcuma xanthorhiza Roxb., Curcuma aromatica Valeton and Zingiber zerumbet Smith (Family: Zingiberaceae) and three synthesized derivatives of xanthorrhizol were evaluated for their ability to inhibit arachidonic acid- (AA), collagen- and
Toxicologic studies of dihydro-5,6-dehydrokawain and 5,6-dehydrokawain.
S Y Hsu et al.
Planta medica, 60(1), 88-90 (1994-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service