MilliporeSigma
All Photos(6)
Key Documents
Z545465
Molymod® Individual organic model set
Protein (beta pleated sheet)
Select a Size
1 SET
$81.20
$81.20
Please contact Customer Service for Availability
Select a Size
Change View
1 SET
$81.20
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
$81.20
Please contact Customer Service for Availability
description
peptides units: 15
manufacturer/tradename
Molymod
General description
The mushroom-shaped hydrogen Atomlink components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45 cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.
Legal Information
Atomlink is a trademark of Spiring Enterprises Ltd.
Molymod is a registered trademark of Spiring Enterprises Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)