Skip to Content
MilliporeSigma
All Photos(1)

Documents

51772

Supelco

(±)-β-Cryptoxanthin

analytical standard

Synonym(s):

(±)-β-Caroten-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56O
CAS Number:
Molecular Weight:
552.87
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

storage temp.

−20°C

General description

β-Cryptoxanthin, a major source of vitamin A, belongs to the class of xanthophyll carotenoids, which show potential activities against chronic, degenerative diseases and also exhibit cancer chemo-preventive property. It is reported to be found in fruits including papayas, mangoes, peaches, oranges and tangerines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cancer chemoprevention by citrus pulp and juices containing high amounts of ?-cryptoxanthin and hesperidin
Tanaka T, et al.
BioMed Research International, 2012 (2011)
Enzymatic formation of apo-carotenoids from the xanthophyll carotenoids lutein, zeaxanthin and ?-cryptoxanthin by ferret carotene-9?, 10?-monooxygenase
Mein RJ, et al.
Archives of Biochemistry and Biophysics, 506(1), 109-121 (2011)
beta-Cryptoxanthin selectively increases in human chylomicrons upon ingestion of tangerine concentrate rich in beta-cryptoxanthin esters
Wingerath T
Archives of Biochemistry and Biophysics, 324(2), 385-390 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service