Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51050

Sigma-Aldrich

Guanosine

≥98.0% (HPLC)

Synonym(s):

9-(β-D-Ribofuranosyl)guanine, Guanine-9-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O5
CAS Number:
Molecular Weight:
283.24
Beilstein:
625911
EC Number:
MDL number:
UNSPSC Code:
41106305

Assay

≥98.0% (HPLC)

optical activity

[α]20/D −73±2°, c = 1.5% in 0.1 M NaOH

impurities

≤3% water

mp

250 °C (dec.) (lit.)

solubility

0.1 M NaOH: 0.1 g/mL, clear, slightly yellow

SMILES string

[C]
[H]O[H].NC1=Nc2c(ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H13N5O5/c11-10-13-7-4(8(19)14-10)12-2-15(7)9-6(18)5(17)3(1-16)20-9/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,19)/t3-,5-,6-,9-/m1/s1

InChI key

NYHBQMYGNKIUIF-UUOKFMHZSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Guo et al.
Journal of natural products, 74(7), 1575-1583 (2011-06-21)
Ten new neo-clerodane diterpenes, ajugaciliatins A-J (1-5, 8-12), along with 17 known analogues (6, 7, 13-27) were isolated from the whole plants of Ajuga ciliata Bunge. Their structures were elucidated by spectroscopic data analysis (IR, ESIMS, HRESIMS, 1D and 2D
Jing Xu et al.
Journal of natural products, 74(10), 2224-2230 (2011-09-21)
Ten myrsinol diterpenes, euphorbiaproliferins A-J (1-10), along with nine known analogues (11-19) were isolated from the roots of Euphorbia prolifera. Their structures were elucidated by spectroscopic data analysis (IR, ESIMS, HRESIMS, 1D and 2D NMR), and the structure of 1
Yanjie Sun et al.
Antimicrobial agents and chemotherapy, 51(1), 84-88 (2006-10-25)
Ribavirin (RBV) is a broad-spectrum antiviral agent that inhibits the production of infectious Hantaan virus (HTNV). Although the mechanism of action of RBV against HTNV is not understood, RBV is metabolized in human cells to both RBV-5'-monophosphate, which inhibits IMP
Craig H Steeves et al.
Bioorganic & medicinal chemistry letters, 21(18), 5188-5190 (2011-08-16)
CTP Synthase from Trypanosoma brucei (TbCTPS) catalyzes the conversion of UTP to CTP and is a recognized target for the development of antiprotozoal agents. GTP activates glutamine-dependent CTP formation catalyzed by TbCTPS at concentrations below 0.2 mM, but inhibits this
Zadkiel Alvarez et al.
Antimicrobial agents and chemotherapy, 54(12), 5329-5336 (2010-10-06)
Bacillus anthracis, the etiological agent of anthrax, has a dormant stage in its life cycle known as the endospore. When conditions become favorable, spores germinate and transform into vegetative bacteria. In inhalational anthrax, the most fatal manifestation of the disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service