MilliporeSigma
  • Bioactive neo-clerodane diterpenoids from the whole plants of Ajuga ciliata Bunge.
Bioactive neo-clerodane diterpenoids from the whole plants of Ajuga ciliata Bunge.
Journal of natural products (2011-06-21)
Ping Guo, Yushan Li, Jing Xu, Cuizhou Liu, Yonggang Ma, Yuanqiang Guo
ABSTRACT
Ten new neo-clerodane diterpenes, ajugaciliatins A-J (1-5, 8-12), along with 17 known analogues (6, 7, 13-27) were isolated from the whole plants of Ajuga ciliata Bunge. Their structures were elucidated by spectroscopic data analysis (IR, ESIMS, HRESIMS, 1D and 2D NMR), and the configuration of 1 was confirmed by X-ray crystallography. All of the compounds were assessed for neuroprotective effects against MPP(+)-induced neuronal cell death in dopaminergic neuroblastoma SH-SY5Y cells. Compounds 2, 6, 7, 9, 10, 15-17, 19, and 20 exhibited moderate neuroprotective effects.
Sigma-Aldrich
Guanosine, ≥97.0% (HPLC)
Sigma-Aldrich
Guanosine, ≥98%
Sigma-Aldrich
Guanosine, BioReagent, suitable for cell culture